Learn More
Biochain Institute Inc Dr. P Kit-Isolation of RNA, DNA and Protein from the same piece of tissue simultaneously, 1 kit/PK
SDP
Catalog No. 50180824
Encompass_Preferred
In order to study the function of DNA, RNA, and protein as well as their relationship, it is very important to get RNA, DNA, and protein from the same piece of tissue. BioChain's Dr.P kit, a patent pending technology, provides a convenient and efficient system for isolation of DNA, RNA, and protein from the same piece of tissue simultaneously. The isolated RNA can be used for mRNA isolation, probe generation, RT-PCR, Northern blot analysis, primer extension, RNA protection assay, and in vitro translation. The isolated DNA can be used for PCR amplification. And the isolated protein can be used for Western Analysis. The kit contains enough reagents for isolating DNA, RNA, and protein from 5 grams of tissue and cells.
Catalog No. 50-180-824 Supplier Biochain Institute Inc Supplier No. K2021010
In order to study the function of DNA, RNA, and protein as well as their relationship, it is very important to get RNA, DNA, and protein from the same piece of tissue. BioChain's Dr.P kit, a patent pending technology, provides a convenient and efficient system for isolation of DNA, RNA, and protein from the same piece of tissue simultaneously. The isolated RNA can be used for mRNA isolation, probe generation, RT-PCR, Northern blot analysis, primer extension, RNA protection assay, and in vitro translation. The isolated DNA can be used for PCR amplification. And the isolated protein can be used for Western Analysis. The kit contains enough reagents for isolating DNA, RNA, and protein from 5 grams of tissue and cells.