Learn More
Marketlab DR Panel Protector GE AMX 700
Catalog No. 22505086
Change view
Click to view available options
Size:
23.69 x 19 in.
Catalog No. Size
22-505-086 23.69 x 19 in.
1 options
Catalog No. 22-505-086 Supplier Marketlab Supplier No. 243009
Add to Cart
Add to Cart
The unique design of the DR Panel Protector provides an easy way to take weight-bearing x-rays of feet (AP view).
  • The unit is simply placed over the DR panel which is first positioned on the floor.
  • Designed to work with most DR systems including those offered by Canon, Siemens, GE Healthcare, Philips, Shimadzu, Quantum, and Trixell.
  • X-ray lucent faceplate
  • High-impact-resistant polycarbonate faceplate
  • 750 lb. weight capacity
  • Nonslip rubber floor grips
  • Nonporous polyethylene and polycarbonate are easy to clean
  • Frame is notched to accommodate the cable connection from the digital DR system to the host panel
Specifications
Product Type Panel Protector
Size 23.69 x 19 in.