Learn More
Aviva Systems Biology Corp DR1 Antibody - N-terminal region (ARP38085_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38085_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DR1. Alternate names: NC2, NC2-BETAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56513