Learn More
Hach Company DR1300 FL Fluorometer 4x AA Batteries Sample Vial Adapter Sample Vial Adapter Cover User Manual
Catalog No. 502253065 Shop All Hach Company Products
Encompass
The DR1300 FL features new fluorescence technology only from Hach. Think about lighting bugs in a night sky. That is what happens when you activate the glow with 10 easy drops of our specially formulated reagent. Then you will ll follow the fluorescence to process improvements by understanding how much chlorine is in your system. - Handheld Fluorometer with innovative fluorescence technology for ultra low range free and total chlorine and sulfite testing Free Chlorine - 2 - 100 ug/L Total Chlorine - 3 - 100 ug/L Sulfite - 6 - 500 ug/L. - The DR1300 FL is suitable for indoor and outdoor use. - Please note that Reagents are not included and have to be purchased separately.
This item is not returnable. View return policy
Catalog No. 50-225-3065 Supplier Hach Company Supplier No. LPV449.97.01002
The DR1300 FL features new fluorescence technology only from Hach. Think about lighting bugs in a night sky. That is what happens when you activate the glow with 10 easy drops of our specially formulated reagent. Then you will ll follow the fluorescence to process improvements by understanding how much chlorine is in your system. - Handheld Fluorometer with innovative fluorescence technology for ultra low range free and total chlorine and sulfite testing Free Chlorine - 2 - 100 ug/L Total Chlorine - 3 - 100 ug/L Sulfite - 6 - 500 ug/L. - The DR1300 FL is suitable for indoor and outdoor use. - Please note that Reagents are not included and have to be purchased separately.