Learn More
ACROBiosystems ACROBiosystems Cynomolgus DR3 / TNFRSF25 Protein, Fc Tag
Catalog No. 501957526
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus DR3, Fc Tag (TN5-C5257) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 203 (Accession # G7NTB5-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-7526 Supplier ACROBiosystems Supplier No. TN5C5257100UG
Recombinant Protein;100UG;Cynomolgus DR3, Fc Tag (TN5-C5257) is expressed from human 293 cells (HEK293). It contains AA Gln 20 - Gly 203 (Accession # G7NTB5-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 50 kDa under reducing (R) condition (SDS-PAGE).