Learn More
Sino Biological Human TNFRSF25 / DR3 / TNFRSF12 Protein (Fc Tag), Biotinylated 20ug
Supplier:  Sino Biological 12095H02HB20
Encompass_Preferred
A DNA sequence encoding the human TNFRSF25 (NP_683867.1) (Met1-Gln199) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5611