Learn More
BPS Bioscience Inc DR3:TL1A Inhibitor Screening Assay Kit, Size: 96 reactions
SDP
Catalog No. NC3221585
Encompass
The DR3:TL1A Inhibitor Screening Assay Kit is designed for screening and profiling inhibitors of the interaction between DR3 (Death Receptor 3) and TL1A (TNF-like ligand 1A). This kit comes in a convenient 96-well or 384-well format, with biotin-labeled TL1A, purified DR3, streptavidin-labeled HRP, and assay buffer for 100 or 400 binding reactions.
Catalog No. NC3221585 Supplier BPS Bioscience Inc Supplier No. 822411
The DR3:TL1A Inhibitor Screening Assay Kit is designed for screening and profiling inhibitors of the interaction between DR3 (Death Receptor 3) and TL1A (TNF-like ligand 1A). This kit comes in a convenient 96-well or 384-well format, with biotin-labeled TL1A, purified DR3, streptavidin-labeled HRP, and assay buffer for 100 or 400 binding reactions.