Learn More
Hach Company DR300 Pocket Colorimeter, Ammonia Nitrogen, with Box
Catalog No. 501997817 Shop All Hach Company Products
Encompass
The DR300 maintains the Pocket Colorimeter legacy of reliability while providing state-of-the-art data transfer capability and connection to Claros*. Reduce data collection hassles, eliminate transcription errors, and ensure stronger compliance traceability.Please note that reagents are not included in the kit and need to be purchased separately.For a limited time the DR300 comes with a FREE 90-day trial of Claros Collect software, allowing measurement data to immediately be available online to your entire team. Learn more at hach.com/claros/InstrumentConnect.*Claros connectivity currently available only in US, Canada and EU.
Catalog No. 50-199-7817 Supplier Hach Company Supplier No. LPV445.97.40110
The DR300 maintains the Pocket Colorimeter legacy of reliability while providing state-of-the-art data transfer capability and connection to Claros*. Reduce data collection hassles, eliminate transcription errors, and ensure stronger compliance traceability.Please note that reagents are not included in the kit and need to be purchased separately.For a limited time the DR300 comes with a FREE 90-day trial of Claros Collect software, allowing measurement data to immediately be available online to your entire team. Learn more at hach.com/claros/InstrumentConnect.*Claros connectivity currently available only in US, Canada and EU.