Learn More
AdipoGen DR6 (mouse):Fc (human) (rec.) (3 x 50 µg)
SDP
Supplier:  AdipoGen AG40B00623050
Encompass_Preferred
Protein. Mouse DR6 (aa 42-348) is fused at the C-terminus to the Fc portion of human IgG1. Source: HEK 293 cells. Endotoxin content: <0.1EU/µg purified protein (LAL test; Lonza). Lyophilized. Contains PBS. Purity: >95% (SDS-PAGE). DR6 is an orphan TNF receptor superfamily member belonging to a subgroup of receptors called death receptors. Expressed ubiquitously with high expression in lymphoid organs, heart, brain, and pancreas. Broadly expressed by developing neurons where it functions as pro-apoptotic factor. Recently, it has been reported that interaction with the beta-amyloid precursor protein (APP) activates a widespread caspase-dependent self-destruction program dependent on caspase-6.
Catalog No. 50-205-3122