Learn More
ACROBiosystems Recombinant Protein;Human DR6 / TNFRSF21 Protein, Fc Tag;HEK293;1MG;TN1,TNFRSF21,DR6,CD358,TNFRSF-21,DR-6,BM-018,BM018
Supplier:  ACROBiosystems TN1H52521MG
Encompass_Preferred
Recombinant Protein;Human DR6 / TNFRSF21 Protein, Fc Tag;1MG;Human DR6, Fc Tag (TN1-H5252) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Leu 350 (Accession # AAH17730).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.2 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8354