Learn More
Medchemexpress LLC Dracorhodin perchlorate | 125536-25-6 | 366.75 | 1 ML
SDP
Catalog No. 5000416656
Encompass_Preferred
Dracorhodin perchlorate is a natural product derived from Dragon's blood. It acts as a pro-apoptotic compound by inhibiting PI3K/Akt and NF-κB activation, upregulating p53 expression, activating caspase, and producing Reactive Oxygen Species (ROS). It also regulates TLR4.
  • Promotes wound healing
  • Improves diabetes
  • Exhibits anti-tumor activity against various cancers including prostate, breast, and cervical cancer
  • Inhibits PI3K/Akt and NF-κB activation
  • Upregulates p53 expression
  • Activates caspase
  • Produces Reactive Oxygen Species (ROS)
  • Regulates TLR4
Catalog No. 50-004-16656 Supplier Medchemexpress LLC Supplier No. HYN072610MM/1ML
Dracorhodin perchlorate is a natural product derived from Dragon's blood. It acts as a pro-apoptotic compound by inhibiting PI3K/Akt and NF-κB activation, upregulating p53 expression, activating caspase, and producing Reactive Oxygen Species (ROS). It also regulates TLR4.
  • Promotes wound healing
  • Improves diabetes
  • Exhibits anti-tumor activity against various cancers including prostate, breast, and cervical cancer
  • Inhibits PI3K/Akt and NF-κB activation
  • Upregulates p53 expression
  • Activates caspase
  • Produces Reactive Oxygen Species (ROS)
  • Regulates TLR4