Learn More
Cayman Chemical Dracorhodin Perchlorate, Purity: ≥98%, CAS: 125536-25-6, 250 mg
SDP
Catalog No. 502940713
Encompass_Preferred
A pro-apoptotic compound and a synthetic analog of dracorhodin; cytotoxic to U937, A375-S2, and MCF-7 cells in a concentration-dependent manner; cytotoxic to HeLa cells, an effect that can be reversed by Ac-YVAD-CMK, A-DEVD-FMK, Z-IETD-FMK, and Z-LEHD-FMK, which are inhibitors of caspase-1, -3, -8, and -9, respectively
Catalog No. 50-294-0713 Supplier Cayman Chemical Supplier No. 27294250 MG
A pro-apoptotic compound and a synthetic analog of dracorhodin; cytotoxic to U937, A375-S2, and MCF-7 cells in a concentration-dependent manner; cytotoxic to HeLa cells, an effect that can be reversed by Ac-YVAD-CMK, A-DEVD-FMK, Z-IETD-FMK, and Z-LEHD-FMK, which are inhibitors of caspase-1, -3, -8, and -9, respectively