Learn More
TARGETMOL CHEMICALS INC DRACORHODIN PERCHLORATE 25MG
SDP
Catalog No. 502580435
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Dracorhodin perchlorate inhibits cell growth and induces apoptosis in fibroblasts in a dose-and time-dependent manner arresting cell cycle at G1 phase may as a candidate for anti-breast cancer. Dracorhodin perchlorate can inhibit high glucose-induced serum and glucocorticoid induced protein kinase 1 (SGK1) and fibronectin(FN) expression in human mesangial cells and this may be part of the mechanism of preventing and treating renal fibrosis of DN. purity: 98%
Catalog No. 50-258-0435 Supplier TARGETMOL CHEMICALS INC Supplier No. T2S204325MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Dracorhodin perchlorate inhibits cell growth and induces apoptosis in fibroblasts in a dose-and time-dependent manner arresting cell cycle at G1 phase may as a candidate for anti-breast cancer. Dracorhodin perchlorate can inhibit high glucose-induced serum and glucocorticoid induced protein kinase 1 (SGK1) and fibronectin(FN) expression in human mesangial cells and this may be part of the mechanism of preventing and treating renal fibrosis of DN. purity: 98%