Learn More
Medchemexpress LLC HY-N0726 5mg Medchemexpress, Dracorhodin perchlorate CAS:125536-25-6 Purity:>98%
Supplier: Medchemexpress LLC HYN07265MG
Medchemexpress, HY-N0726 5mg Dracorhodin perchlorate CAS:125536-25-6 Dracorhodin perchlorate (Dracohodin perochlorate) is a natural product extracted from a natural medicine Dragon's blood. Dracorhodin perchlorate inhibits cell proliferation and induces cell cycle arrest and apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.