Learn More
Draeger Safety Inc Draeger MicroTubes, Sulfur Dioxide, 0.05 to 5 ppm
Catalog No. NC3975515
Encompass_Preferred
MicroTubes are an innovative analysis system component for precise, on-site measurement of gases, reducing the need for lab analyses. Each set offers 10 individual tests using substance-specific reactive and pre-layers to filter out interfering substances, ensuring accurate target gas indication and minimizing cross-sensitivities. An integrated RFID tag provides one year of valid calibration data, removing manual calibration needs and lowering ownership costs. This specific MicroTube detects Sulfur Dioxide, 0.05 to 5 ppm.
  • Precise gas measurement.
  • Immediate, on-site results.
  • 10 tests per set.
  • Substance-specific selective testing.
  • Filters interfering substances.
  • RFID tag with one-year calibration data.
  • Eliminates manual calibration; reduces costs.
  • Laboratory-quality results with portability.
Catalog No. NC3975515 Supplier Draeger Safety Inc Supplier No. 8610110
MicroTubes are an innovative analysis system component for precise, on-site measurement of gases, reducing the need for lab analyses. Each set offers 10 individual tests using substance-specific reactive and pre-layers to filter out interfering substances, ensuring accurate target gas indication and minimizing cross-sensitivities. An integrated RFID tag provides one year of valid calibration data, removing manual calibration needs and lowering ownership costs. This specific MicroTube detects Sulfur Dioxide, 0.05 to 5 ppm.
  • Precise gas measurement.
  • Immediate, on-site results.
  • 10 tests per set.
  • Substance-specific selective testing.
  • Filters interfering substances.
  • RFID tag with one-year calibration data.
  • Eliminates manual calibration; reduces costs.
  • Laboratory-quality results with portability.