Learn More
Mettler Toledo™ Draft Shield Magic Cube XSR
Catalog No. 01919113 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
XPR/XSR Balances with 1 mg Readability
Catalog No. For Use With (Equipment)
01-919-113 XPR/XSR Balances with 1 mg Readability
1 options
Catalog No. 01-919-113 Supplier Mettler Toledo™ Supplier No. 30300928
Add to Cart
Add to Cart
Protects weighing samples from surrounding influences by minimizing air turbulence and ensuring fast stabilization
  • Draft Shield Magic Cube XSR
  • Fast Stabilization Times
  • Compatible with XPR/XSR Balances with readability of 1 mg
  • Protects weighing samples from surrounding influences
  • Minimizing air turbulence
  • Ensures faster stabilization
  • Shield can be removed in a few seconds for quick and effortless cleaning
  • Height: 175 mm
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XPR/XSR Balances with 1 mg Readability
Height (Metric) 175 mm
For Use With (Application) To Protect Weighing Samples From Surrounding Influences
Type Draft Shield for Balances
Autoclavable Not Autoclavable