Learn More
Mettler Toledo™ Draft Shield Pro XPR
Catalog No. 01919112 Shop All METTLER TOLEDO Products
Change view
Click to view available options
248 mm
Catalog No.
01-919-112 248 mm
1 options
Catalog No. 01-919-112 Supplier Mettler Toledo™ Supplier No. 30300926
Add to Cart
Add to Cart
Protects weighing samples from surrounding influences by minimizing air turbulence and ensuring fast stabilization
  • Draft Shield Pro XPR
  • Fast Stabilization Times
  • Compatible with XPR/XSR Balances with a readability of 1 mg/0.1 mg
  • Patented draft shield doors do not slide open beyond the balance
  • Easily removed in a few seconds for quick, effortless cleaning
  • Saves valuable space within laboratories
  • Protects weighing samples from surrounding influences
  • Minimizes air turbulence
  • Ensures faster stabilization
  • Height: 248 mm
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XPR/XSR Balances with 1 mg/0.1 mg Readability
Height (Metric) 248 mm
For Use With (Application) To Protect Weighing Samples From Surrounding Influences
Type Draft Shield for Balances
Autoclavable Not Autoclavable