Learn More
SPT LABTECH INC Dragonfly Discovery Sterile Reservoirs, 50/pk
Catalog No. NC1818883
Encompass
Individual and disposable polypropylene reservoirs, each with a 10 mL capacity and 300 uL fully recoverable dead volume. The reservoirs load directly into the instrument reservoir tray and help ensure easy loading of assay reagents, with fill level indicators for 5 mL and 10 mL levels. Contains 50 gamma ray sterilised reservoirs.
Catalog No. NC1818883 Supplier SPT LABTECH INC Supplier No. 415007204
Individual and disposable polypropylene reservoirs, each with a 10 mL capacity and 300 uL fully recoverable dead volume. The reservoirs load directly into the instrument reservoir tray and help ensure easy loading of assay reagents, with fill level indicators for 5 mL and 10 mL levels. Contains 50 gamma ray sterilised reservoirs.