Learn More
SPT LABTECH INC Dragonfly Discovery Ultra Low Retention Syringes | 100/pk
Catalog No. NC1818905
Encompass
These unique disposable, non-contact, positive-displacement syringes feature an external hydrophobic/ oleophobic coating to dispense concentrated detergent or protein solutions. With a volume range of 200 nL to 4 mL they are suitable for dispensing into 96-1536 well microplates.
Catalog No. NC1818905 Supplier SPT LABTECH INC Supplier No. 415007208
These unique disposable, non-contact, positive-displacement syringes feature an external hydrophobic/ oleophobic coating to dispense concentrated detergent or protein solutions. With a volume range of 200 nL to 4 mL they are suitable for dispensing into 96-1536 well microplates.