Learn More
New England Biolabs, Inc. DraI – 10000 units
SDP
Catalog No. 50811976
Encompass
A restriction endonuclease that recognizes the sequence TTT_^AAA.
Catalog No. 50-811-976 Supplier New England Biolabs, Inc. Supplier No. R0129L
A restriction endonuclease that recognizes the sequence TTT_^AAA.