Learn More
New England Biolabs, Inc. DraI – 2000 units
SDP
Catalog No. 50811977
Encompass
A restriction endonuclease that recognizes the sequence TTT_^AAA.
Catalog No. 50-811-977 Supplier New England Biolabs, Inc. Supplier No. R0129S
A restriction endonuclease that recognizes the sequence TTT_^AAA.