Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BUCHI Drain Disk
SDP

Catalog No. 05405029 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Rotavapor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-405-029 Rotavapor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-405-029 Supplier BUCHI Supplier No. 040625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Drain disk is constructed of PTFE with an inner diameter of 5.1 mm, and an outer diameter of 14 mm.

  • Constructed of PTFE
  • Inner Diameter: 5.1 mm
  • Outer Diameter: 14 mm

Specifications

Material PTFE
Type Drain Disk
For Use With (Application) Solvent Feeding Hose
For Use With (Equipment) Rotavapor
Diameter (Metric) 5.1 mm (Inner), 14 mm (Outer)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.