Learn More
MSC Detco Liquid Alkaline Drain Opener, 1 Quart
Catalog No. 502816777
Encompass
Clean Line is an alkaline drain opener designed to keep pipes free flowing and prevent stoppages. It effectively liquefies grease and dissolves hair and paper without containing harmful acids or producing fumes. This product contains potassium hydroxide and should not be mixed with other drain openers.
  • Dissolves fats, oils, grease and hair
  • Safe on pipes and septic systems
  • Fast, penetrating action
  • Made in the U.S.A.
  • Lab tested
  • Industry approved
  • Biodegradable
  • Safe and effective, guaranteed
Catalog No. 50-281-6777 Supplier MSC Supplier No. 33686528
Clean Line is an alkaline drain opener designed to keep pipes free flowing and prevent stoppages. It effectively liquefies grease and dissolves hair and paper without containing harmful acids or producing fumes. This product contains potassium hydroxide and should not be mixed with other drain openers.
  • Dissolves fats, oils, grease and hair
  • Safe on pipes and septic systems
  • Fast, penetrating action
  • Made in the U.S.A.
  • Lab tested
  • Industry approved
  • Biodegradable
  • Safe and effective, guaranteed