Learn More
Med Vet International Silicone Wound Drain, 10/Box
Catalog No. 502830101
Encompass_Preferred
Traditional Style 100% Silicone Wound Drains Provide Effective Drainage. Smooth, heat polished perforations are positioned in alternate 90 degree planes for effective exudates drainage. Drain placement is quick and easy thanks to the convenient indicator dot. Available in various round and flat sizes. Radiopaque. Double-Wrapped Sterile Package. Round Silicone Drains 19Fr, 1/4" End-Perforation.
Catalog No. 50-283-0101 Supplier Med Vet International Supplier No. DYNJWE1325A
Traditional Style 100% Silicone Wound Drains Provide Effective Drainage. Smooth, heat polished perforations are positioned in alternate 90 degree planes for effective exudates drainage. Drain placement is quick and easy thanks to the convenient indicator dot. Available in various round and flat sizes. Radiopaque. Double-Wrapped Sterile Package. Round Silicone Drains 19Fr, 1/4" End-Perforation.