Learn More
Med Vet International Silicone Thoracic Drain, 24fr. 8.0mm, J0524B
Catalog No. 5000495077
Encompass_Preferred
Straight with X-ray opaque line with an open end. Graduated depth markings. Six smooth oval lateral eyes. Autoclavable. 50cm L.
Catalog No. 50-004-95077 Supplier Med Vet International Supplier No. JOR524B
Straight with X-ray opaque line with an open end. Graduated depth markings. Six smooth oval lateral eyes. Autoclavable. 50cm L.