Learn More
Med Vet International Silicone Thoracic Drain, 16fr. 5.3mm, J0524
Catalog No. 5000495084
Encompass_Preferred
Straight with X-ray opaque line with an open end. Graduated depth markings. Six smooth oval lateral eyes. Autoclavable. 50cm L.
Catalog No. 50-004-95084 Supplier Med Vet International Supplier No. JOR524
Straight with X-ray opaque line with an open end. Graduated depth markings. Six smooth oval lateral eyes. Autoclavable. 50cm L.