Learn More
Med Vet International Closed Wound Drainage Kit, Wound Kit w/Flat Drain, Silicone, 100cc, J0790B, 1 Kit
Catalog No. 5000495597
Encompass_Preferred
This vacuum drain‘s most common use is for post-op drainage Jackson Pratt. The kit is designed to create suction or negative pressure for draining wounds. A fenestrated silicone drain is placed into position with the aid of a stainless steel trocar. This drain is attached to a silicone bulb with a one-way valve. When the bulb is released a vacuum is created that pulls exudates out through the drain. Wound exudates can be stored, visualized, or cultured. The drain has a radio-opaque line for the entire length.The kit consists of: a 130mm stainless steel trocar, silicone fenestrated drain and tubing, a silicone bulb and a clamp for attachment to bandage or patient.
Catalog No. 50-004-95597 Supplier Med Vet International Supplier No. JOR790B
This vacuum drain‘s most common use is for post-op drainage Jackson Pratt. The kit is designed to create suction or negative pressure for draining wounds. A fenestrated silicone drain is placed into position with the aid of a stainless steel trocar. This drain is attached to a silicone bulb with a one-way valve. When the bulb is released a vacuum is created that pulls exudates out through the drain. Wound exudates can be stored, visualized, or cultured. The drain has a radio-opaque line for the entire length.The kit consists of: a 130mm stainless steel trocar, silicone fenestrated drain and tubing, a silicone bulb and a clamp for attachment to bandage or patient.