Learn More
Med Vet International Silicone Chest Drainage Tube, J0524C, 12Fr, 4mm x 2mm, Each
Catalog No. 5000495582
Encompass_Preferred
Multifenestration with black numbered markings at 2 and 4cm from proximal hole. It has an opened end. White color. Proximal end can be trimmed to desired length.
Catalog No. 50-004-95582 Supplier Med Vet International Supplier No. JOR524C
Multifenestration with black numbered markings at 2 and 4cm from proximal hole. It has an opened end. White color. Proximal end can be trimmed to desired length.