Learn More
Julabo USA Inc Drain tap with 8 mm ID tube for PURA/SW water baths.
SDP
Catalog No. 501904635
Encompass_Preferred
Drain tap with 8 mm ID tube for JULABO PURA / SW baths. Threads onto drain port with manual valve for easy draining. Suitable for use with PURA 10, 14, 22, 30 and SW22, SW23.
Catalog No. 50-190-4635 Supplier Julabo USA Inc Supplier No. 8970453
Drain tap with 8 mm ID tube for JULABO PURA / SW baths. Threads onto drain port with manual valve for easy draining. Suitable for use with PURA 10, 14, 22, 30 and SW22, SW23.