Learn More
Med Vet International McKesson Wound Drain with Trocar, 10 Fr., 12-Inch Length, 370017, Each
Catalog No. 503044660
Encompass_Preferred
Wound Drain Tube McKesson Brand Silicone Perforated Style 10 Fr. Size Sterile Discrete drains contain internal ribs to help prevent collapse and occlusion of the drain 10-millimeter discrete drain offers minimal transition from the drain hub to the drain tubing for ease of removal and added patient comfort Radiopaque Sterile, round type, silicone wound drain tube is 10 Fr.
Catalog No. 50-304-4660 Supplier Med Vet International Supplier No. 491312EA
Wound Drain Tube McKesson Brand Silicone Perforated Style 10 Fr. Size Sterile Discrete drains contain internal ribs to help prevent collapse and occlusion of the drain 10-millimeter discrete drain offers minimal transition from the drain hub to the drain tubing for ease of removal and added patient comfort Radiopaque Sterile, round type, silicone wound drain tube is 10 Fr.