Learn More
Perkin Elmer US LLC Drain Tubing for NexION ICP-MS Instruments
Catalog No. 502104398
Encompass_Preferred
The tubing has a 0.038 in inner diameter and a 0.062 in outer diameter. Note that this tubing should be inspected and replaced on a reoccurring basis as part of a regular maintenance schedule.
Catalog No. 50-210-4398 Supplier Perkin Elmer US LLC Supplier No. 02506495
The tubing has a 0.038 in inner diameter and a 0.062 in outer diameter. Note that this tubing should be inspected and replaced on a reoccurring basis as part of a regular maintenance schedule.