Learn More
Perkin Elmer US LLC Drain Tubing for S10/AS-93plus
Catalog No. 502117302
Encompass_Preferred
The tubing has a 6.4 mm inner diameter, a 1.6 mm wall thickness and is clear in color. Note that the tubing is part of the Rinsing Kit B3140236.
Catalog No. 50-211-7302 Supplier Perkin Elmer US LLC Supplier No. B0509650
The tubing has a 6.4 mm inner diameter, a 1.6 mm wall thickness and is clear in color. Note that the tubing is part of the Rinsing Kit B3140236.