Learn More
Shimadzu Scientific Instruments DRAIN VALVE ASSEMBLY SHIMADZU
Catalog No. NC3667756
Encompass
Drain Valve Assembly LC-20AD, LC-2030, LC-40DDrain valve assembly for the following pumps: LC-20AD, LC-20AB, LC-2030, LC-40D. Not for LC-40XR/XS/X3. Replaces 228-45574-91
Catalog No. NC3667756 Supplier Shimadzu Scientific Instruments Supplier No. 2284557495
Drain Valve Assembly LC-20AD, LC-2030, LC-40DDrain valve assembly for the following pumps: LC-20AD, LC-20AB, LC-2030, LC-40D. Not for LC-40XR/XS/X3. Replaces 228-45574-91