Learn More
Aviva Systems Biology Corp DRAM Antibody - N-terminal region (ARP58891_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58891_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DRAM. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DRAM. Alternate names: DRAM, FLJ11259ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-517