Learn More
Med Vet International Sterile Surgical Drape, 40-in x 58-in
Catalog No. 502522401
Encompass_Preferred
Sterile surgical drapes made of SMS non-woven material. Consists of two outer layers of SMS and a middle layer of Outer layers: Cloth-like, yet tough and abrasion-resistant spunbound polypropylene. Inner layer: Intricate web of microfibers filter out many water-based liquids and dry particles.
Catalog No. 50-252-2401 Supplier Med Vet International Supplier No. DR4058
Sterile surgical drapes made of SMS non-woven material. Consists of two outer layers of SMS and a middle layer of Outer layers: Cloth-like, yet tough and abrasion-resistant spunbound polypropylene. Inner layer: Intricate web of microfibers filter out many water-based liquids and dry particles.