Learn More
Med Vet International Eent Drape 3M Steri-Drape Small Drape With Aperture 15 W X 15 L inch Sterile, 1020, Each
Catalog No. 502830893
Encompass_Preferred
Small drape with adhesive aperture is made of impervious polyethylene film and provides a barrier to liquids, reducing exposure to blood and body fluids Circular aperture has adhesive film that isolates the surgical site from lid margins, eyelashes and eyebrows to help reduce the risk of surgical site contamination Matte finish helps reduce glare from drape surface during procedure 15 x 15 inches
Catalog No. 50-283-0893 Supplier Med Vet International Supplier No. 5717EA
Small drape with adhesive aperture is made of impervious polyethylene film and provides a barrier to liquids, reducing exposure to blood and body fluids Circular aperture has adhesive film that isolates the surgical site from lid margins, eyelashes and eyebrows to help reduce the risk of surgical site contamination Matte finish helps reduce glare from drape surface during procedure 15 x 15 inches