Learn More
Med Vet International Surgical Drape 3M Steri-Drape Aperture Drape 9-1/4 W X 18 L inch Sterile, 1072, 40/Case
Catalog No. 502829482
Encompass_Preferred
Fluid collection pouch of the 1072 drape helps minimize the risk of blood and body fluid exposure to healthcare personnel, and eliminates need to add pouch to the drape, saving time and money Reliable and secure adhesive
Catalog No. 50-282-9482 Supplier Med Vet International Supplier No. 5725CS
Fluid collection pouch of the 1072 drape helps minimize the risk of blood and body fluid exposure to healthcare personnel, and eliminates need to add pouch to the drape, saving time and money Reliable and secure adhesive