Learn More
Med Vet International Orthopedic Drape 3M Steri-Drape Arthroscopy Drape With Pouch 89 W X 125 L inch Sterile, 1194, 5/Case
Catalog No. 502831103
Encompass_Preferred
Two elastomeric apertures isolate the operative site at the knee and the ankle Low profile roomy pouch stays out of the surgeon’s way Dual exit ports on both sides of rectangular pouch provide better fluid control Malleable metal wire keeps the pouch open for optimal fluid collection
Catalog No. 50-283-1103 Supplier Med Vet International Supplier No. 191833CS
Two elastomeric apertures isolate the operative site at the knee and the ankle Low profile roomy pouch stays out of the surgeon’s way Dual exit ports on both sides of rectangular pouch provide better fluid control Malleable metal wire keeps the pouch open for optimal fluid collection