Learn More
Med Vet International Fenestrated Drape, 40"x40", 7"x1.25", Rectangle, Each
Catalog No. 502522537
Encompass_Preferred
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.
Catalog No. 50-252-2537 Supplier Med Vet International Supplier No. 40407
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.