Learn More
Med Vet International Autoclavable Fenestrated Drape, 40" X 40", 10"X 1.25" Rectangle, Each
Catalog No. 502831199
Encompass_Preferred
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.
Catalog No. 50-283-1199 Supplier Med Vet International Supplier No. FD4010
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.