Learn More
Med Vet International Fenestrated Drape, 40"X40", 10"X1-1/4", 10/Case
Catalog No. 502829075
Encompass_Preferred
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.
Catalog No. 50-282-9075 Supplier Med Vet International Supplier No. 404010CS
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.