Learn More
Med Vet International Autoclavable Fenestrated Drape, 40" X 40", 7" X 1.25" Rectangle, Each
Catalog No. 502830816
Encompass_Preferred
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.
Catalog No. 50-283-0816 Supplier Med Vet International Supplier No. FD407
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.