Learn More
Med Vet International Fenestrated Drape, 20"x20", 4"x3/4", Each
Catalog No. 502522573
Encompass_Preferred
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.
Catalog No. 50-252-2573 Supplier Med Vet International Supplier No. 20204
Cloth fenestrated drapes made from high quality surgical sheeting with built-in moisture barrier. Drapes are colorfast, washable, autoclavable and come in a choice of sizes.