Learn More
Med Vet International Autoclavable Fenestrated Drape, 30" X 30", 2" X 1" Rectangle, Each
Catalog No. 502830338
Encompass_Preferred
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.
Catalog No. 50-283-0338 Supplier Med Vet International Supplier No. FD302
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.