Learn More
Med Vet International Autoclavable Fenestrated Drape, 30" X 30", 7" X 1.25" Rectangle, Each
Catalog No. 502829029
Encompass_Preferred
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.
Catalog No. 50-282-9029 Supplier Med Vet International Supplier No. FD307
Cloth fenestrated drape made from high-quality surgical sheeting. Colorfast, washable, autoclavable and size may vary approximately 1", no choice of color.