Learn More
Med Vet International 3M Ioban 2 Sterile Antimicrobial Incise Surgical Drape, 17 X 23in, 6650Ez, 10/Box
Catalog No. 502830189
Encompass_Preferred
Surgical Drape 3M™Ioban™2 Antimicrobial Incise Drape 17 W X 23 L Inch Sterile Polyethylene liner, full-width handle, and imprinted strip at one end Liner comes off smoothly, effortlessly, with no tearing, for easier drape delivery Handle controls liner release and drape application Strip is imprinted with stop signs to indicate when the end of the drape is near Surgical drapes with an antimicrobial film designed to provide a sterile surface all the way to the wound edge and continuous antimicrobial activity throughout the procedure Incise drapes are made with a "low memory" film that conforms to body contours and stretches to allow limb manipulation Yellow surgical drape is 17 x 23 inches
Catalog No. 50-283-0189 Supplier Med Vet International Supplier No. 401845BX
Surgical Drape 3M™Ioban™2 Antimicrobial Incise Drape 17 W X 23 L Inch Sterile Polyethylene liner, full-width handle, and imprinted strip at one end Liner comes off smoothly, effortlessly, with no tearing, for easier drape delivery Handle controls liner release and drape application Strip is imprinted with stop signs to indicate when the end of the drape is near Surgical drapes with an antimicrobial film designed to provide a sterile surface all the way to the wound edge and continuous antimicrobial activity throughout the procedure Incise drapes are made with a "low memory" film that conforms to body contours and stretches to allow limb manipulation Yellow surgical drape is 17 x 23 inches