Learn More
Med Vet International Surgical Drape 3M Steri-Drape Large Pouch With Incise Film 30 W X 30 L inch Sterile, 6658, 20/Case
Catalog No. 502831108
Encompass_Preferred
Full-circumference surgical drape pouches that collect blood and body fluids to help reduce the risk of contamination 3M Ioban 2 antimicrobial incise film provides continuous broad-spectrum antimicrobial activity to meet your procedural needs 360 degree fluid control pouch is rectangle in shape There are twenty 30 x 30 inch surgical drapes in a case
Catalog No. 50-283-1108 Supplier Med Vet International Supplier No. 177163CS
Full-circumference surgical drape pouches that collect blood and body fluids to help reduce the risk of contamination 3M Ioban 2 antimicrobial incise film provides continuous broad-spectrum antimicrobial activity to meet your procedural needs 360 degree fluid control pouch is rectangle in shape There are twenty 30 x 30 inch surgical drapes in a case