Learn More
Med Vet International 3M Ioban 2 Sterile Antimicrobial Incise Surgical Drape, 23 X 33in, 6651Ez, Each
Catalog No. 502829025
Encompass_Preferred
Surgical Drape 3M™Ioban™2 Antimicrobial Incise Drape 23 W X 33 L Inch Sterile Polyethylene liner, full-width handle, and imprinted strip at one end Liner comes off smoothly, effortlessly, with no tearing, for easier drape delivery Handle controls liner release and drape application Strip is imprinted with stop signs to indicate when the end of the drape is near Surgical drapes with an antimicrobial film designed to provide a sterile surface all the way to the wound edge and continuous antimicrobial activity throughout the procedure Incise drapes are made with a "low memory" film that conforms to body contours and stretches to allow limb manipulation 23 inches wide x 33 inches long
Catalog No. 50-282-9025 Supplier Med Vet International Supplier No. 404185EA
Surgical Drape 3M™Ioban™2 Antimicrobial Incise Drape 23 W X 33 L Inch Sterile Polyethylene liner, full-width handle, and imprinted strip at one end Liner comes off smoothly, effortlessly, with no tearing, for easier drape delivery Handle controls liner release and drape application Strip is imprinted with stop signs to indicate when the end of the drape is near Surgical drapes with an antimicrobial film designed to provide a sterile surface all the way to the wound edge and continuous antimicrobial activity throughout the procedure Incise drapes are made with a "low memory" film that conforms to body contours and stretches to allow limb manipulation 23 inches wide x 33 inches long