Learn More
Med Vet International Surgical Drape 3M Steri-Drape Large Incise Drape 17 W X 23 L inch Sterile, 1050, Each
Catalog No. 502830927
Encompass_Preferred
Drape creates a sterile surface at the beginning of surgery Drape adhesive adheres securely to skin and immobilizes bacteria Plastic film provides a barrier to liquids Incise draping maximizes OR efficiency by providing fluid control Saves time and money by helping to eliminate the need to add pouch to the drape Aperture drapes have adjustable apertures that allow for custom fitting to each patient, helping to decrease inventory and cost of drapes 17 inches wide x 23 inches long There are 40 clear Steri-Drape incise drapes in a case
Catalog No. 50-283-0927 Supplier Med Vet International Supplier No. 5721EA
Drape creates a sterile surface at the beginning of surgery Drape adhesive adheres securely to skin and immobilizes bacteria Plastic film provides a barrier to liquids Incise draping maximizes OR efficiency by providing fluid control Saves time and money by helping to eliminate the need to add pouch to the drape Aperture drapes have adjustable apertures that allow for custom fitting to each patient, helping to decrease inventory and cost of drapes 17 inches wide x 23 inches long There are 40 clear Steri-Drape incise drapes in a case