Learn More
Med Vet International Surgical Drape 3M Steri-Drape Irrigation Pouch 19 W X 23 L inch Sterile, 1016, 10/Box
Catalog No. 502830346
Encompass_Preferred
Irrigation pouches with an adhesive strip that are designed to collect irrigation fluids during surgical and minor procedures Translucent matte-finished film allows for visualization of fluids in pouch while reducing glare from drape surface 19 inches wide x 23 inches long
Catalog No. 50-283-0346 Supplier Med Vet International Supplier No. 113533BX
Irrigation pouches with an adhesive strip that are designed to collect irrigation fluids during surgical and minor procedures Translucent matte-finished film allows for visualization of fluids in pouch while reducing glare from drape surface 19 inches wide x 23 inches long